Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 3 mol/l (3 N) in aqueous solution analytical reagent
Corrosive
Sodium hydroxide 3 mol/l (3 N) in aqueous solution analytical reagent
Catalog # 47124.K2
CAS Number:  
Corrosive
Sodium hydroxide 3 mol/l (3 N) in aqueous solution analytical reagent
Catalog # 47124.K2
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548