Order Entry
Belgium
ContactUsLinkComponent
Viscosity standard, Specpure®, nominally 50cSt @ 40° and 7.3cSt @ 100°
Viscosity standard, Specpure®, nominally 50cSt @ 40° and 7.3cSt @ 100°
Catalog # 47013.AP
CAS Number:  
Viscosity standard, Specpure®, nominally 50cSt @ 40° and 7.3cSt @ 100°
Catalog # 47013.AP
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    Viscosity standard, Nominally 50cSt at 40 and 7.3cSt at 100°
  • MDL No.:
    MFCD00084411
  • Physical form:
    Liquid
  • Odour/scent:
    Odorless

Specifications

About this item

Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00084411