Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Petroleum Industry Standards
Specpure® Sulfur in Isooctane Standard Solution
Specpure® Sulfur in Isooctane Standard Solution
Specpure® Sulfur in Isooctane Standard Solution
Catalog # 45353.AE
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Expand image
Specpure® Sulfur in Isooctane Standard Solution
Catalog # 45353.AE
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Certificates
Specifications
Pk:
100 mL
Description:
Sulfur in Isooctane standard solution 20 µg/g (0.0020%)
Specifications
About this item
Product Details
Documentation
Product Details
Documentation