Order Entry
Belgium
ContactUsLinkComponent
Viscosity standard, Specpure®, Nominal Values at 40 and 100 °C
Viscosity standard, Specpure®, Nominal Values at 40 and 100 °C
Certificates
Viscosity standard, Specpure®, Nominal Values at 40 and 100 °C
Certificates

Specifications

  • MDL No.:
    MFCD00084411
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless

Specifications

Related Information

Frequently Bought Together