Lead in isooctane standard solution, Specpure®, 37 µg/g(0,1 g/gal)
Molecular Formula: Matrix: Isooctane Physical Form: Liquid Odour: Nearly Odourless UN No.: UN1262 MDL No.: MFCD01094200
- MDL No.:MFCD01094200
- UN No.:1262
- Physical form:Liquid
- Odour/scent:Nearly Odourless
- Molecular formula:Matrix: Isooctane
- Appearance:Colourless
Frequently Bought Together