Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cesium nitrate, ionization buffer solution, Specpure®

Cesium nitrate, ionization buffer solution, Specpure®

Cesium Nitrate Buffer solution, 100 mL
Catalog # 45294.AE
Cesium nitrate, ionization buffer solution, Specpure®
Catalog # 45294.AE
CAS Number:  
Cesium nitrate, ionization buffer solution, Specpure®
Cesium nitrate, ionization buffer solution, Specpure®

Cesium Nitrate Buffer solution, 100 mL

CAS Number:  
Price
130,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

CAS No.: 7789-18-6
Molecular Formula: 1% Cs (as Cs₂CO₃) in 5% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00010963

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    Cesium Nitrate Buffer solution
  • UN No.:
    3264
  • Cat. No.:
    45294.AE
  • Molecular formula:
    1% Cs (as Cs2CO3) in 5% HNO3
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00010963
  • CAS No.:
    7789-18-6

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.