Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Palladium nitrate and magnesium nitrate, matrix modifier solution, Specpure®

Palladium nitrate and magnesium nitrate, matrix modifier solution, 2% HNO₃, 250 mL
  45291.AK
Palladium nitrate and magnesium nitrate, matrix modifier solution, Specpure®
  45291.AK
 :  
Palladium nitrate and magnesium nitrate, matrix modifier solution, Specpure®
Palladium nitrate and magnesium nitrate, matrix modifier solution, Specpure®

Palladium nitrate and magnesium nitrate, matrix modifier solution, 2% HNO₃, 250 mL

 :  
 
212,00 €
   
1 * 250 mL
 

Pre-mixed matrix modifier are designed for use with graphite furnace atomic absorption (GFAA).

UN No.: UN3264
MDL No.: MFCD09910543

 

 

  • Matrix:
    2% HNO₃
  • Pk:
    250 mL
  • Description:
    Palladium nitrate and magnesium nitrate, matrix modifier solution
  • Cat. No.:
    45291.AK
  • MDL No.:
    MFCD09910543