Order Entry
Belgium
ContactUsLinkComponent
Palladium nitrate and magnesium nitrate, matrix modifier solution, Specpure®
Palladium nitrate and magnesium nitrate, matrix modifier solution, Specpure®
Catalog # 45290.AK
CAS Number:  
Palladium nitrate and magnesium nitrate, matrix modifier solution, Specpure®
Catalog # 45290.AK
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pk:
    250 mL
  • Description:
    Palladium nitrate & magnesium nitrate, matrix modifier solution
  • MDL No.:
    MFCD09910543

Specifications

About this item

Pre-mixed matrix modifier are designed for use with graphite furnace atomic absorption (GFAA).

UN No.: UN3264
MDL No.: MFCD09910543