Selenium 78, 78 Se, plasma standard solution, 78 Se 10 µg/ml, Specpure®
45283.AD
:
- Concentration:10 mg/L
- Matrix:2% HNO₃
- Pk:50 mL
- Environmentally Preferable:
- MDL No.:MFCD00084579
- Element:Selenium
- Symbol:Se
Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00084579