Order Entry
Belgium
ContactUsLinkComponent
Lithium 6, 6 Li, plasma standard solution, 6 Li 100 µg/ml, Specpure®
Lithium 6, 6 Li, plasma standard solution, 6 Li 100 µg/ml, Specpure®
Catalog # 45280.AE
CAS Number:  
Lithium 6, 6 Li, plasma standard solution, 6 Li 100 µg/ml, Specpure®
Catalog # 45280.AE
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • MDL No.:
    MFCD00084621
  • Element:
    Lithium
  • Symbol:
    Li

Specifications

About this item

Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00084621