Order Entry
Belgium
ContactUsLinkComponent
Iron 57, ⁵⁷Fe, plasma standard solution, Specpure®
Iron 57, ⁵⁷Fe, plasma standard solution, Specpure®
Catalog # 45277.AD
CAS Number:  
Iron 57, ⁵⁷Fe, plasma standard solution, Specpure®
Catalog # 45277.AD
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    50 mL
  • MDL No.:
    MFCD00144503
  • Element:
    Iron
  • Symbol:
    Fe

Specifications

About this item

UN No.: UN3264
MDL No.: MFCD00144503

Caution: 10 mg/l = 10 µg/ml=10 ppm