Order Entry
Belgium
ContactUsLinkComponent
 
Chromium 50, 50 Cr, plasma standard solution, 50 Cr 10 µg/ml, Specpure®
  45275.AD
 :  
Chromium 50, 50 Cr, plasma standard solution, 50 Cr 10 µg/ml, Specpure®
  45275.AD
 :  

 

  • Concentration:
    10 µg/g
  • Matrix:
    2% HNO₃
  • Pk:
    50 mL
  • MDL No.:
    MFCD00190543
  • Element:
    Chromium
  • Symbol:
    Cr

 

 

Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00190543