
Dichloro[bis(diphenylphosphinophenyl)ether]palladium(II) (14.8% Pd)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:205319-06-8
:
Formula:C₃₆H₂₈Cl₂OP₂Pd
MDL Number:MFCD09953446
:715.887 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together