Bis(tri-o-tolylphosphine)palladium(II) dichloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:40691-33-6
:
Formula:C₄₂H₄₂Cl₂P₂Pd
MDL Number:MFCD00274659
:786.07 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together