
[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride (17.6% Pd)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:29964-62-3
:
Formula:C₂₈H₂₈Cl₂P₂Pd
MDL Number:MFCD02093437
:603.80 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together