Order Entry
Belgium
ContactUsLinkComponent
Instrument calibration(ICAL) standard, Solution B, Specpure®
Instrument calibration(ICAL) standard, Solution B, Specpure®
Catalog # 44856.AE
CAS Number:  
Instrument calibration(ICAL) standard, Solution B, Specpure®
Catalog # 44856.AE
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Instrument Calibration(ICAL) Standard, Solution B
  • MDL No.:
    MFCD08064527

Specifications

About this item

Physical Form: Liquid
UN No.: UN1789
MDL No.: MFCD08064527

Concentration:
Al 1000 µg/ml ±2%
As 1000 µg/ml ±2%
Ca 10,000 µg/ml ±2%
Cr 1000 µg/ml ±2%
K 10,000 µg/ml ±2%
Mg 10,000 µg/ml ±2%
Na 10,000 µg/ml ±2%
Sb 1000 µg/ml ±2%
Se 1000 µg/ml ±2%
V 1000 µg/ml ±2%