Order Entry
Belgium
ContactUsLinkComponent
 
(Perfluorocyclohexyl)methyl acrylate ≥96% stabilised
Catalog # 44793.06
CAS Number:  
(Perfluorocyclohexyl)methyl acrylate ≥96% stabilised
Catalog # 44793.06
CAS Number:  

Specifications

  • Pk:
    5 g

Specifications

About this item

 :
 :366.13 g/mol
 :Stabilised with 4-methoxyphenol (MEHQ) 50 ppm-100 ppm
Storage Temperature:Refrigerator
CAS Number:40677-94-9
Formula:C₁₀H₅F₁₁O₂
MDL Number:MFCD00080614