Order Entry
Belgium
ContactUsLinkComponent
ICP-MS interference check standard solution B for 6020, Specpure®
ICP-MS interference check standard solution B for 6020, Specpure®
Catalog # 44713.AE
CAS Number:  
ICP-MS interference check standard solution B for 6020, Specpure®
Catalog # 44713.AE
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    100 mL
  • Description:
    ICP-MS Interference Check Standard solution B for 6020, Specpure®
  • MDL No.:
    MFCD00803324

Specifications

About this item

Matrix: 5% HNO₃
UN No.: UN3264
MDL No.: MFCD00803324

Note: To be diluted 1:500 to achieve prescribed concentrations.

Concentration: Ag 10 µg/ml ±2%; Al 10 µg/ml ±2%; As 10 µg/ml ±2%; Cd 10 µg/ml ±2%; Co 10 µg/ml ±2%; Cr 10 µg/ml ±2%; Cu 10 µg/ml ±2%; Mn 10 µg/ml ±2%; Ni 10 µg/ml ±2%; Se 10 µg/ml ±2%; V 10 µg/ml ±2%; Zn 10 µg/ml ±2%