Alkali metals, plasma standard solution, Ba,Be,Ca,Cs,K,Li,Mg,Na,Rb,Sr @ 100 µg/ml, Specpure®
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
UN No.: UN3264
MDL No.: MFCD08061357
Spécifications
- MDL No.:MFCD08061357
Spécifications
Frequently Bought Together