Order Entry
Belgium
ContactUsLinkComponent
ICP-MS internal standard for 6020, Specpure®
ICP-MS internal standard for 6020, Specpure®
Catalog # 43161.AE
CAS Number:  
ICP-MS internal standard for 6020, Specpure®
Catalog # 43161.AE
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    ICP-MS Internal Standard for 6020, Specpure®
  • MDL No.:
    MFCD01868829

Specifications

About this item

Molecular Formula: Matrix: 5% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD01868829