Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Multi-element plasma standard solution 3, Specpure®

Multi-Element Plasma Standard Solution 3, Nitric acid dil., 100 mL
Catalog # 42884.AE
Multi-element plasma standard solution 3, Specpure®
Catalog # 42884.AE
CAS Number:  
Multi-element plasma standard solution 3, Specpure®
Multi-element plasma standard solution 3, Specpure®

Multi-Element Plasma Standard Solution 3, Nitric acid dil., 100 mL

CAS Number:  
Price
312,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

UN No.: UN3264
MDL No.: MFCD00212339

Specifications

Specifications

  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • Description:
    Multi-Element Plasma Standard Solution 3
  • Cat. No.:
    42884.AE
  • MDL No.:
    MFCD00212339

Specifications