Order Entry
Belgium
ContactUsLinkComponent
ICP-MS soil spike standard solution for 6020 CLP-M, Specpure®
ICP-MS soil spike standard solution for 6020 CLP-M, Specpure®
Catalog # 42604.AE
CAS Number:  
ICP-MS soil spike standard solution for 6020 CLP-M, Specpure®
Catalog # 42604.AE
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃/tr. HF
  • Pk:
    100 ml
  • Description:
    ICP-MS soil spike standard solution for 6020 CLP-M
  • MDL No.:
    MFCD02266211

Specifications

About this item

Molecular Formula: Matrix: 2% HNO₃/tr. tartaric acid
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD02266211