Order Entry
Belgium
ContactUsLinkComponent
 
ICP-MS calibration standard solution for 6020 CLP-M, Specpure®
Catalog # 42602.AE
CAS Number:  
ICP-MS calibration standard solution for 6020 CLP-M, Specpure®
Catalog # 42602.AE
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃/tr. HF
  • Pk:
    100 mL
  • Description:
    ICP-MS calibration standard solution for 6020 CLP-M
  • MDL No.:
    MFCD03792349

Specifications

About this item

UN No.: UN3264
MDL No.: MFCD03792349