Order Entry
Belgium
ContactUsLinkComponent
ICP-MS stock standard solution A for 200,8, Rev. 5,4, Specpure®
ICP-MS stock standard solution A for 200,8, Rev. 5,4, Specpure®
Catalog # 42596.AE
CAS Number:  
ICP-MS stock standard solution A for 200,8, Rev. 5,4, Specpure®
Catalog # 42596.AE
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃/tr. (CHOH)₂(COOH)₂
  • Pk:
    100 mL
  • Description:
    ICP-MS Stock Standard Solution A for 200.8, Rev. 5.4
  • MDL No.:
    MFCD03792352

Specifications

About this item

UN No.: UN3264
MDL No.: MFCD03792352