Order Entry
Belgium
ContactUsLinkComponent
ICP-MS tuning standard solution for HP®, Specpure®
ICP-MS tuning standard solution for HP®, Specpure®
Catalog # 42586.AE
CAS Number:  
ICP-MS tuning standard solution for HP®, Specpure®
Catalog # 42586.AE
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    100 mL
  • Description:
    ICP-MS Tuning Standard Solution for HP®
  • MDL No.:
    MFCD00240453

Specifications

About this item

UN No.: UN3264
MDL No.: MFCD00240453