Specpure® Iodide, Ion Chromatography Standard Solution, I- 1000 µg/ml
Supplier: Thermo Fisher Scientific
Certificates
About this item
Specifications
- MDL No.:MFCD00011355
- Physical form:Liquid
- Molecular formula:NaI in H2O
Specifications
Related Information
Frequently Bought Together