Hydrazine, standard solution, Specpure®, N₂H₄ 100 µg/ml
Catalog # 42561.AO
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Matrix:1% CH₃COOH
- Pk:450 mL
- Description:Hydrazine standard
- MDL No.:MFCD00011417
- Formula weight:50.06
- Physical form:Liquid
- Odour/scent:Ammonia-like
Specifications
About this item
Molecular Formula: N₂H₄ in 1% CH₃CO₂H
Formula Weight: 50,06
Physical Form: Liquid
Odour: Ammonia-like
MDL No.: MFCD00011417