Order Entry
Belgium
ContactUsLinkComponent
Multi Ion cation IC standard solution in 5% HNO₃, Specpure® (Dionex)
Multi Ion cation IC standard solution in 5% HNO₃, Specpure® (Dionex)
Catalog # 41695.AE
CAS Number:  
Multi Ion cation IC standard solution in 5% HNO₃, Specpure® (Dionex)
Catalog # 41695.AE
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    100 mL
  • Description:
    Multi Ion Cation IC Standard Solution
  • Environmentally Preferable:
  • MDL No.:
    MFCD03792456

Specifications

About this item

Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD03792456