Order Entry
Belgium
ContactUsLinkComponent
 
Multi Ion anion IC standard solution in H₂O, Specpure®
Catalog # 41693.AE
CAS Number:  
Multi Ion anion IC standard solution in H₂O, Specpure®
Catalog # 41693.AE
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Multi Ion anion IC standard solution in H₂O, Specpure®
  • MDL No.:
    MFCD00144814
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless

Specifications

About this item

Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00144814