Nitrite, Ion chromatography standard solution, Specpure®, NO₂- 1000 µg/ml
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: NaNO2 in H2O
Physical Form: Liquid
MDL No.: MFCD00144923
Specifications
- MDL No.:MFCD00144923
- Physical form:Liquid
- Molecular formula:NaNO2 in H2O
Specifications
Related Information
Frequently Bought Together