Order Entry
Belgium
ContactUsLinkComponent
Ground & Waste Water Pollution Standard Solution 3, Specpure®
Ground & Waste Water Pollution Standard Solution 3, Specpure®
Catalog # 36750.AE
CAS Number:  
Ground & Waste Water Pollution Standard Solution 3, Specpure®
Catalog # 36750.AE
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Description:
    Ground & Waste Water Pollution Standard Solution 3
  • MDL No.:
    MFCD02091710
  • UN No.:
    3264

Specifications

About this item

For US EPA Clean Water Act.

  • 6 Analytes

UN No.: 3264
MDL No.: MFCD02091710