Order Entry
Belgium
ContactUsLinkComponent
Ground & Waste Water Pollution Standard Solution 2, Specpure®
Ground & Waste Water Pollution Standard Solution 2, Specpure®
Certificates
Ground & Waste Water Pollution Standard Solution 2, Specpure®

Specifications

  • MDL No.:
    MFCD02091712
  • UN No.:
    3264

Specifications

Related Information

Frequently Bought Together