Order Entry
Belgium
ContactUsLinkComponent
 
Multi Ion anion IC standard solution, Specpure® (Waters) in H₂O
Catalog # 35564.AE
CAS Number:  
Multi Ion anion IC standard solution, Specpure® (Waters) in H₂O
Catalog # 35564.AE
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Multi Ion anion IC standard solution, Specpure® (Waters) in H₂O
  • MDL No.:
    MFCD00144814
  • Physical form:
    Liquid

Specifications

About this item

Physical Form: Liquid
MDL No.: MFCD00144814