

Palladium(II) chloride solution (9.0 - 11.0% Pd) (w/w)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-10-1
:
Formula:PdCl₂
MDL Number:MFCD00003558
ADR:8,II
:177.33 g/mol
EINECS:231-596-2
UN:3264
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together