

Bis(acetonitrile)palladium(II) dichloride (40.5% Pd)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14592-56-4
:
Formula:C₄H₆Cl₂N₂Pd
MDL Number:MFCD00013122
:259.43 g/mol
EINECS:238-637-3
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together