Order Entry
Belgium
ContactUsLinkComponent
Dissolved oxygen sensor, Ox 1113T
Dissolved oxygen sensor, Ox 1113T
Catalog # SCOI285206410
Supplier:  SI Analytics
Dissolved oxygen sensor, Ox 1113T
Catalog # SCOI285206410
Supplier:  SI Analytics

Specifications

  • Connection:
    1 m fixed cable with 8-pole plug
  • Ø×L:
    12×120 mm
  • Type:
    Ox 1113T
  • Description:
    DO sensor, amperometric, Ox 1113T
  • Material:
    Plastic
  • Pk:
    1
  • Temp.:
    −5...+45 °C

Specifications

About this item

Maintenance set

Product Information