Anti-RAB7A Rabbit Polyclonal Antibody
Catalog # BSBTPB9883
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:RAB7A, member RAS oncogene family
- Antigen symbol:RAB7A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7879
- Antigen synonyms:PRO2706|RAB7
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: RAB7A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat