Anti-CXCL2 Rabbit Polyclonal Antibody
Catalog # BSBTPB9858
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-X-C motif) ligand 2
- Antigen symbol:CXCL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:114105
- Antigen synonyms:Mip-2|Scyb2
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CXCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat