Anti-TGFBR1 Rabbit Polyclonal Antibody
100 µg
Catalog # BSBTPB9828
Supplier: Boster Bio
Jump to:
Product Details & Documents
Type: Primary
Antigen: TGFBR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Documents
PB9828
Datasheet
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Isotype:IgG
- Host:Rabbit
- Cross adsorption:No
- Antigen name:transforming growth factor, beta receptor 1
- Cat. No.:BSBTPB9828
- Reactivity:Human
- Gene ID:7046
- Antigen synonyms:tbetaR-I|LDS1|LDS1A|MSSE|LDS2A|AAT5|ACVRLK4|ALK-5|SKR4|ESS1|ALK5|TGFR-1
- Conjugation:Unconjugated
- Antigen symbol:TGFBR1



