Anti-SULT2A1 Rabbit Polyclonal Antibody
Catalog # BSBTPB9821
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1
- Antigen symbol:SULT2A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6822
- Antigen synonyms:hSTa|ST2|ST2A3|DHEAS|STD|ST2A1|HST|DHEA-ST
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SULT2A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat