Anti-STXBP2 Rabbit Polyclonal Antibody
BSBTPB9820
- Antibody type:Primary
- Antigen name:syntaxin binding protein 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6813
- Antigen synonyms:MUNC18-2|UNC18B|Hunc18b|FHL5|UNC18-2|pp10122
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: STXBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat