Order Entry
Belgium
ContactUsLinkComponent
BD Vacutainer® SPC Plus blood collection tubes with BD Hemogard™ closures
BD Vacutainer® SPC Plus blood collection tubes with BD Hemogard™ closures
Catalog # BDAM368380
Supplier:  BD Medical
BD Vacutainer® SPC Plus blood collection tubes with BD Hemogard™ closures
Catalog # BDAM368380
Supplier:  BD Medical

Specifications

  • Closure type:
    BD Hemogard™
  • Ø:
    13 mm
  • Volume:
    6 ml
  • Description:
    Blood Collection tube, BD Vacutainer®
  • Length:
    100 mm
  • Material:
    Plastic
  • Cap colour:
    Royal blue
  • Pk:
    100
  • Additive:
    Silica, acts as Serum clot activator

Specifications

About this item