Orders LinkContactUsLinkComponent
Capillary dispenser, VITREX®
Capillary dispenser, VITREX®
Capillary tube dispenser for use with most Vitrex capillary tubes, Grey
Catalog # MODU569013
Supplier:   Vitrex Medical A/S
Capillary dispenser, VITREX®
Catalog # MODU569013
Supplier:  Vitrex Medical A/S
Capillary dispenser, VITREX®
Capillary dispenser, VITREX®
Capillary tube dispenser for use with most Vitrex capillary tubes, Grey
Price
48,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
This capillary dispenser is a unique liquid handling tool, developed to empty capillary tubes for blood or other liquids.
  • Two step plunger operation facilitates super blow-out from the tip
  • Its unique tip cone is designed to accept most VITREX capillary tubes
  • At a length of 130 mm this pipette is the optimum practical size for blood collecting
  • Fully autoclavable at 121 °C & 15 psi
This dispenser is specially designed for use with Diagnostic Kits.
Specifications
  • Colour code:
    Grey
  • Description:
    Capillary tube dispenser for use with most Vitrex capillary tubes