Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cobalt standard solution

Cobalt standard solution

Cobalt standard solution, 100 ml (NIST), 1000 mg/l as Co, 100 mL
Catalog # 705-2029
Supplier:   Hach
Cobalt standard solution
Catalog # 705-2029
Supplier:  Hach
CAS Number:  
Cobalt standard solution
Cobalt standard solution

Cobalt standard solution, 100 ml (NIST), 1000 mg/l as Co, 100 mL

CAS Number:  
Price
46,20 €
Log in for availability
1 * 100 mL
Product Details & Documents

Standard solution for accuracy checks in cobalt determinations.

Specifications

Specifications

  • Concentration:
    1000 mg/l as Co
  • Pk:
    100 mL
  • Description:
    Cobalt standard solution, 100 ml (NIST)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.