Orders LinkContactUsLinkComponent
Test kits, chloride, Spectroquant®, Supelco®
Test kits, chloride, Spectroquant®, Supelco®
Test kits, chloride, Spectroquant®, Supelco®
Supplier:  Merck
Certificates
Test kits, chloride, Spectroquant®, Supelco®
Test kits, chloride, Spectroquant®, Supelco®
4 Options Available Below
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1.01807.0001
  • 1.01804.0001
  • 1.14897.0001
  • 1.14730.0001
  • M-Clarity quality level:
  • MQ100
  • MQ100
Specifications
Product Family Options
Product Information
  • Range
    Chemical method
    For unit
    Pk
    Availability
    Price
  • 1.01807.0001
    Chloride test
    Chloride test
    0,10 - 5,00 mg/L Cl
    Iron (III) thiocyanate
    Prove, Nova 60, Multy, Move
    100 Tests
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 186,00 €
    186,00 €
    1 * 1 EA
       186,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.01807.0001
    M-Clarity quality level
  • 1.01804.0001
    Chloride cell test
    Chloride cell test
    0,5 - 15,0 mg/L Cl
    Iron (III) thiocyanate
    Prove, Nova 60, Multy, Move
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 130,00 €
    130,00 €
    1 * 25 Tests
       130,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.01804.0001
    M-Clarity quality level
  • Chloride test
    2,5 - 250 mg/L Cl
    Iron (III) thiocyanate
    Prove, Nova 60, Multy, Move
    100 Tests
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA - 188,00 €
    188,00 €
    1 * 1 EA
       188,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.14897.0001
    M-Clarity quality level
    MQ100
  • Chloride cell test
    5 - 125 mg/L Cl
    Iron (III) thiocyanate
    Prove, Nova 30/60, Multy, Move
    25 Tests
    Log in for availability
    Log in for availability
    1 * 25 Tests
    1 * 25 Tests - 147,00 €
    147,00 €
    1 * 25 Tests
       147,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    1.14730.0001
    M-Clarity quality level
    MQ100