ContactUsLinkComponent
Quartz microfibre filters for air sampling, grade QM-B, Whatman™
Quartz microfibre filters for air sampling, grade QM-B, Whatman™
Certificates
undefined
Quartz microfibre filters for air sampling, grade QM-B, Whatman™
Specifications
  • Catalog No:
  • WHAT1852-042
  • WHAT1852-040
  • Weight:
  • 170 g/m²
  • 170 g/m²
  • Thickness:
  • 950 µm
  • 950 µm
  • Airflow rate:
  • 12 s/100 ml/6,5 cm²
  • 12 s/100 ml/6,5 cm²
  • Max. temperature:
  • 800 °C
  • 800 °C