ContactUsLinkComponent
Test kits, hydrazine, Spectroquant®, Supelco®
Test kits, hydrazine, Spectroquant®, Supelco®
Catalog # 1.09711.0001
Supplier:  Merck
CAS Number:  
undefined
Test kits, hydrazine, Spectroquant®, Supelco®
Catalog # 1.09711.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Chemical method:
    4-(Dimethylamino)benzaldehyde
  • Pk:
    100 Tests
  • Description:
    Hydrazine test
  • For unit:
    Prove, Nova 60, Multy, Move
  • Range:
    0,005 - 2,00 mg/l N₂H₄
Specifications
About this item
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60 A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Highly stable, ready to use reagent mixtures in a user friendly design
  • Free selection of appropriate cell format according to the measuring range
  • Long shelf life up to 3 years - all at room temperature
  • Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.