Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Bottle Closures
Screw caps for bottles
Screw caps for bottles
Screw caps for bottles
Catalog # WITG5.039.702
Supplier:
witeg Labortechnik
Expand image
Screw caps for bottles
Catalog # WITG5.039.702
Supplier:
witeg Labortechnik
Specifications
Thread:
GL 40
Colour:
White
Description:
Screw cap
Seq:
02
Pk:
100
Specifications
About this item
Product Details
Product Details