Anti-NCR1 Rat Monoclonal Antibody (Biotin) [clone: 29A1.4]
Supplier: BIOLEGEND INC
Certificates
About this item
Type: Primary
Antigen: NCR1
Clonality: Monoclonal
Clone: 29A1.4
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Specifications
- Catalog No:
- BLEG137615
- BLEG137616
- Antigen symbol:
- NCR1
- NCR1
- Antigen name:
- Natural cytotoxicity triggering receptor 1
- Natural cytotoxicity triggering receptor 1
- Antigen synonyms:
- CD335|NK-p46|LY94|NKP46
- CD335|NK-p46|LY94|NKP46
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Biotin
- Biotin
- Clone:
- 29A1.4
- 29A1.4
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 17086
- 17086
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- NKP46-IgG1 Fc fusion protein
- NKP46-IgG1 Fc fusion protein
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with biotin under optimal conditions. The solution is free of unconjugated biotin.
- The antibody was purified by affinity chromatography, and conjugated with biotin under optimal conditions. The solution is free of unconjugated biotin.
- Concentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Storage temperature:
- The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together