Orders LinkContactUsLinkComponent
Membrane filters, type 184
Membrane filters, type 184
   Sustainable Options Available
Supplier:   Sartorius
Membrane filters, type 184
Supplier:  Sartorius
   Sustainable Options Available
Certificates
Membrane filters, type 184
12 Options Available Below
Product Details & Documents
Regenerated cellulose hydrophilic membrane filters. For filtration of organic solvents. Suitable for particle removal from aqueous solutions and solvents. Low adsorption.
* Sterile
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 512-6116
  • 512-6216
  • 512-6316
  • 512-6416
  • SART18406-50-ACN
  • 512-6516
  • 512-6118
  • 512-6218
  • 512-6318
  • 512-6418
  • SART18407-142-N
  • 512-6518
  • Type:
  • 184
  • 184
  • 184
  • 184
  • 184
  • 184
  • 184
  • 184
  • 184
  • 184
  • 184
  • 184
  • Pore size:
  • 0,45 µm
  • 0,45 µm
  • 0,45 µm
  • 0,45 µm
  • 0,45 µm
  • 0,45 µm
  • 0,20 µm
  • 0,20 µm
  • 0,20 µm
  • 0,20 µm
  • 0,20 µm
  • 0,20 µm
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Thickness:
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • Water flow rate:
  • 30 ml/min/cm² (1 bar)
  • 30 ml/min/cm² (1 bar)
  • 30 ml/min/cm² (1 bar)
  • 30 ml/min/cm² (1 bar)
  • 30 ml/min/cm² (1 bar)
  • 30 ml/min/cm² (1 bar)
  • 15 ml/min/cm² (1 bar)
  • 15 ml/min/cm² (1 bar)
  • 15 ml/min/cm² (1 bar)
  • 15 ml/min/cm² (1 bar)
  • 15 ml/min/cm² (1 bar)
  • 15 ml/min/cm² (1 bar)
  • Max. temperature:
  • 180 °C
  • 180 °C
  • 180 °C
  • 180 °C
  • 180 °C
  • 180 °C
  • 180 °C
  • 180 °C
  • 180 °C
  • 180 °C
  • 180 °C
  • 180 °C
  • Environmental certification:
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
  • Renewable Energy Certificate
Specifications
Product Family Options
Product Information
  • Type
    Pore size
    Ø
    Sterile
    Availability
    Price
  • 184
    0,45 µm
    13 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 158,00 €
    158,00 €
    1 * 100 ST
       158,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    512-6116
    Type
    184
    Pore size
    0,45 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    170 µm
    Water flow rate
    30 ml/min/cm² (1 bar)
    Max. temperature
    180 °C
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 184
    0,45 µm
    25 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 179,00 €
    179,00 €
    1 * 100 ST
       179,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    512-6216
    Type
    184
    Pore size
    0,45 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    170 µm
    Water flow rate
    30 ml/min/cm² (1 bar)
    Max. temperature
    180 °C
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 184
    0,45 µm
    47 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 266,00 €
    266,00 €
    1 * 100 ST
       266,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    512-6316
    Type
    184
    Pore size
    0,45 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    170 µm
    Water flow rate
    30 ml/min/cm² (1 bar)
    Max. temperature
    180 °C
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 184
    0,45 µm
    50 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 265,00 €
    265,00 €
    1 * 100 ST
       265,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    512-6416
    Type
    184
    Pore size
    0,45 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    170 µm
    Water flow rate
    30 ml/min/cm² (1 bar)
    Max. temperature
    180 °C
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 184
    0,45 µm
    50 mm
    +
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 309,00 €
    309,00 €
    1 * 100 ST
       309,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SART18406-50-ACN
    Type
    184
    Pore size
    0,45 µm
    Environmentally Preferable
    Thickness
    170 µm
    Water flow rate
    30 ml/min/cm² (1 bar)
    Max. temperature
    180 °C
    Environmental certification
  • 184
    0,45 µm
    100 mm
    -
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 182,00 €
    182,00 €
    1 * 25 ST
       182,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    512-6516
    Type
    184
    Pore size
    0,45 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    170 µm
    Water flow rate
    30 ml/min/cm² (1 bar)
    Max. temperature
    180 °C
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 184
    0,20 µm
    13 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 156,00 €
    156,00 €
    1 * 100 ST
       156,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    512-6118
    Type
    184
    Pore size
    0,20 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    170 µm
    Water flow rate
    15 ml/min/cm² (1 bar)
    Max. temperature
    180 °C
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 184
    0,20 µm
    25 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 181,00 €
    181,00 €
    1 * 100 ST
       181,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    512-6218
    Type
    184
    Pore size
    0,20 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    170 µm
    Water flow rate
    15 ml/min/cm² (1 bar)
    Max. temperature
    180 °C
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 184
    0,20 µm
    47 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 260,00 €
    260,00 €
    1 * 100 ST
       260,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    512-6318
    Type
    184
    Pore size
    0,20 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    170 µm
    Water flow rate
    15 ml/min/cm² (1 bar)
    Max. temperature
    180 °C
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 184
    0,20 µm
    50 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 265,00 €
    265,00 €
    1 * 100 ST
       265,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    512-6418
    Type
    184
    Pore size
    0,20 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    170 µm
    Water flow rate
    15 ml/min/cm² (1 bar)
    Max. temperature
    180 °C
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 184
    0,20 µm
    142 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 1 420,00 €
    1 420,00 €
    1 * 100 ST
       1 420,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    SART18407-142-N
    Type
    184
    Pore size
    0,20 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    170 µm
    Water flow rate
    15 ml/min/cm² (1 bar)
    Max. temperature
    180 °C
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact
  • 184
    0,20 µm
    100 mm
    -
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 182,00 €
    182,00 €
    1 * 25 ST
       182,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    512-6518
    Type
    184
    Pore size
    0,20 µm
    Environmentally Preferable
    Low Manufacturing Impact
    Thickness
    170 µm
    Water flow rate
    15 ml/min/cm² (1 bar)
    Max. temperature
    180 °C
    Environmental certification
    Renewable Energy Certificate
      Low Manufacturing Impact